missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boron trifluoride, 12% (1.5M) in methanol, Thermo Scientific Chemicals

Boron trifluoride, 12% (1.5M) in methanol, ACROS Organics, Quantity: 500g, Packaging: Glass bottle, Boiling Point: 65.0 deg.C, CAS: 373-57-9, 67-56-1, Amber to Colorless, Melting Point: -98.0 deg.C, Molecular Formula: BF3, Molecular Weight: 67.81, Percent Purity: 10 to 15% (w/v BF3)

$65.14 - $283.87

Specifications

Boiling Point 65.0°C
Chemical Name or Material Boron trifluoride
Melting Point -98.0°C
Name Note 1.5M solution in methanol
CAS 373-57-9,67-56-1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC402761000
View Documents
Thermo Scientific Chemicals
402761000
100 g Glass bottle
Each for $65.14
 
AC402765000
View Documents
Thermo Scientific Chemicals
402765000
500 g Glass bottle
Each for $160.05
 
AC402760010
View Documents
Thermo Scientific Chemicals
402760010
1 kg Glass bottle
Each for $283.87
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent, Lewis acid
Specifications

Specifications

65.0°C
-98.0°C
373-57-9,67-56-1
Solution
BF3
4°C
boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution
WTEOIRVLGSZEPR-UHFFFAOYSA-N
67.81
67.81
Glass bottle
0.8700g/mL
Boron trifluoride
1.5M solution in methanol
Amber to Colorless
BF3
MFCD00011316
14, 1349
Solubility in water: may decompose
FB(F)F
11062313
10 to 15% (w/v BF3)
0.87
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only