missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boron trifluoride dihydrate, 65% BF3, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 388711000

 View more versions of this product

Catalog No. AC388711000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

58°C to 60°C (1.5mmHg)
6°C
Colorless to Yellow
100 mL
BF3H4O2
MFCD00149901
Solubility in water: miscible
O.O.FB(F)F
103.84
103.85
Confirmed
1.642
Boron trifluoride dihydrate
13319-75-0
Liquid
65.0 to 66%
BF3·2H2O
boron trifluoride dihydrate, boron fluoride dihydrate, borane, trifluoro-, dihydrate, trifluoroborane dihydrate, bf3.2h2o, bf3 dihydrate, bortrifluorid dihydrat, borontrifluoride dihydrate, dsstox_cid_13264, dsstox_rid_79058
MJCYPBSRKLJZTB-UHFFFAOYSA-N
trifluoroborane dihydrate
61591
65% (BF3)
Glass bottle
1.6420g/mL

Chemical Identifiers

13319-75-0
103.84
MJCYPBSRKLJZTB-UHFFFAOYSA-N
61591
O.O.FB(F)F
BF3H4O2
MFCD00149901
boron trifluoride dihydrate, boron fluoride dihydrate, borane, trifluoro-, dihydrate, trifluoroborane dihydrate, bf3.2h2o, bf3 dihydrate, bortrifluorid dihydrat, borontrifluoride dihydrate, dsstox_cid_13264, dsstox_rid_79058
trifluoroborane dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only