Learn More
Boron trifluoride-dimethyl sulfide complex, purified, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
$615.42 - $615.42
Chemical Identifiers
CAS | 353-43-5 |
---|---|
Molecular Formula | C2H6BF3S |
Molecular Weight (g/mol) | 129.94 |
MDL Number | MFCD00013193 |
InChI Key | BRWZPVRDOUWXKE-UHFFFAOYSA-N |
Synonym | boron fluoride-dimethyl sulfide complex, dimethyl-??-sulfanylidene trifluoro-??-borane, trifluoroborane-methyl sulfide, dimethyl sulfide-trifluoroborane, trifluoro methylsulfanyl methane boron, boron trifluoride methyl sulfide complex, boron trifluoride-methyl sulfide complex, boron trifluoride-dimethyl sulfide complex, purified, packaged under argon in resealable chemseal bottles |
PubChem CID | 71311438 |
IUPAC Name | dimethylsulfonio(trifluoro)boranuide |
SMILES | CSC.FB(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA42970AE
|
Thermo Scientific Chemicals
042970AE |
100 mL |
Each for $615.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
353-43-5 | |
129.94 | |
BRWZPVRDOUWXKE-UHFFFAOYSA-N | |
71311438 | |
CSC.FB(F)F |
C2H6BF3S | |
MFCD00013193 | |
boron fluoride-dimethyl sulfide complex, dimethyl-??-sulfanylidene trifluoro-??-borane, trifluoroborane-methyl sulfide, dimethyl sulfide-trifluoroborane, trifluoro methylsulfanyl methane boron, boron trifluoride methyl sulfide complex, boron trifluoride-methyl sulfide complex, boron trifluoride-dimethyl sulfide complex, purified, packaged under argon in resealable chemseal bottles | |
dimethylsulfonio(trifluoro)boranuide |
Specifications
353-43-5 | |
100 mL | |
MFCD00013193 | |
boron fluoride-dimethyl sulfide complex, dimethyl-??-sulfanylidene trifluoro-??-borane, trifluoroborane-methyl sulfide, dimethyl sulfide-trifluoroborane, trifluoro methylsulfanyl methane boron, boron trifluoride methyl sulfide complex, boron trifluoride-methyl sulfide complex, boron trifluoride-dimethyl sulfide complex, purified, packaged under argon in resealable chemseal bottles | |
CSC.FB(F)F | |
129.94 | |
129.94 | |
Moisture sensitive | |
Boron trifluoride-dimethyl sulfide complex, Purified |
Liquid | |
C2H6BF3S | |
UN2920 | |
BRWZPVRDOUWXKE-UHFFFAOYSA-N | |
dimethylsulfonio(trifluoro)boranuide | |
71311438 | |
ChemSeal™ bottles | |
1.235 g/mL |
Safety and Handling
GHS H Statement
H224-H314-H318-H335
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
TSCA : No
storageNote1 :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only