Learn More
Boron trifluoride etherate, ca. 48% BF3, AcroSeal™, Thermo Scientific Chemicals
$144.57 - $309.34
Chemical Identifiers
CAS | 109-63-7 |
---|---|
Molecular Formula | C4H10BF3O |
Molecular Weight (g/mol) | 141.93 |
MDL Number | MFCD00013194 |
InChI Key | KZMGYPLQYOPHEL-UHFFFAOYSA-N |
Synonym | Boron trifluoride ethyl ether |
PubChem CID | 8000 |
IUPAC Name | ethoxyethane;trifluoroborane |
SMILES | FB(F)F.CCOCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC427111000
|
Thermo Scientific Chemicals
427111000 |
100 mL | AcroSeal™ Glass Bottle |
Each for $144.57
|
|
||||
AC427118000
|
Thermo Scientific Chemicals
427118000 |
800 mL | AcroSeal™ Glass Bottle |
Each for $309.34
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
109-63-7 | |
141.93 | |
KZMGYPLQYOPHEL-UHFFFAOYSA-N | |
8000 | |
FB(F)F.CCOCC |
C4H10BF3O | |
MFCD00013194 | |
Boron trifluoride ethyl ether | |
ethoxyethane;trifluoroborane |
Specifications
109-63-7 | |
Colorless to Straw-Yellow | |
126.0°C | |
AcroSeal™ Glass Bottle | |
1.3429 to 1.3449 | |
MFCD00013194 | |
1.12 | |
Boron trifluoride ethyl ether | |
KZMGYPLQYOPHEL-UHFFFAOYSA-N | |
ethoxyethane;trifluoroborane | |
8000 | |
141.93 | |
Liquid |
-60.0°C | |
1.1200g/mL | |
58°C | |
C4H10BF3O | |
(C2H5)2O·BF3 | |
100 mL | |
15, 1352 | |
Solubility in water: reacts. Other solubilities: decomposes in alcohol,soluble in ether | |
FB(F)F.CCOCC | |
141.93 | |
1.89 mPa.s (20°C) | |
ca. 48% BF3 | |
Boron trifluoride etherate, ca. 48% BF3 |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
EINECSNumber : 203-689-8
RUO – Research Use Only