Learn More
Boron trifluoride-tetrahydrofuran complex, BF{3} 45.5%, Thermo Scientific Chemicals
$65.74 - $166.92
Chemical Identifiers
CAS | 462-34-0 |
---|---|
Molecular Formula | C4H8BF3O |
Molecular Weight (g/mol) | 139.91 |
MDL Number | MFCD00040372 |
InChI Key | CHNLPLHJUPMEOI-UHFFFAOYSA-N |
Synonym | boron trifluoride tetrahydrofuran complex, trifluoro tetrahydro-1h-furan-1-ium-1-yl borate, pubchem7062, trifluoro oxolan-1-ium-1-yl boranuide, borontrifluoride tetrahydrofuran complex, boron trifluoride tetrahydrofuran complex, contains sulfur dioxide as peroxide formation suppressor |
PubChem CID | 60146129 |
SMILES | FB(F)F.C1CCOC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL1759322
|
Thermo Scientific Chemicals
L1759322 |
100 g |
Each for $65.74
|
|
||||||
AAL1759336
|
Thermo Scientific Chemicals
L1759336 |
500 g |
Each for $166.92
|
|
||||||
Description
Boron trifluoride-tetrahydrofuran complex is used as a polymerization initiator and as a dopant in the semiconductor industry. It serves as a reagent for deoxofluorination of alcohols and carbonyls. Further, it is used in the synthesis of methacrylate-PEG-methacrylate-boron trifluoride self-doped gel polymer electrolytes and hydroboration-oxidation of divinylferrocene. In addition to this, it is used as a reagent for the preparation of NK1 receptor antagonists, complex polyketide natural products and selective serotonin reuptake inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBoron trifluoride-tetrahydrofuran complex is used as a polymerization initiator and as a dopant in the semiconductor industry. It serves as a reagent for deoxofluorination of alcohols and carbonyls. Further, it is used in the synthesis of methacrylate-PEG-methacrylate-boron trifluoride self-doped gel polymer electrolytes and hydroboration-oxidation of divinylferrocene. In addition to this, it is used as a reagent for the preparation of NK1 receptor antagonists, complex polyketide natural products and selective serotonin reuptake inhibitors.
Solubility
Miscible with dimethyl formamide.
Notes
Incompatible with strong oxidizing agents and water.
Chemical Identifiers
462-34-0 | |
139.91 | |
CHNLPLHJUPMEOI-UHFFFAOYSA-N | |
60146129 |
C4H8BF3O | |
MFCD00040372 | |
boron trifluoride tetrahydrofuran complex, trifluoro tetrahydro-1h-furan-1-ium-1-yl borate, pubchem7062, trifluoro oxolan-1-ium-1-yl boranuide, borontrifluoride tetrahydrofuran complex, boron trifluoride tetrahydrofuran complex, contains sulfur dioxide as peroxide formation suppressor | |
FB(F)F.C1CCOC1 |
Specifications
462-34-0 | |
1.268 | |
92°C (197°F) | |
MFCD00040372 | |
UN2922 | |
boron trifluoride tetrahydrofuran complex, trifluoro tetrahydro-1h-furan-1-ium-1-yl borate, pubchem7062, trifluoro oxolan-1-ium-1-yl boranuide, borontrifluoride tetrahydrofuran complex, boron trifluoride tetrahydrofuran complex, contains sulfur dioxide as peroxide formation suppressor | |
CHNLPLHJUPMEOI-UHFFFAOYSA-N | |
oxolane; trifluoroborane | |
60146129 | |
Boron trifluoride-tetrahydrofuran complex |
8°C to 9°C | |
180°C | |
C4H8BF3O | |
100 g | |
Moisture sensitive | |
Miscible with dimethyl formamide. | |
FB(F)F.C1CCOC1 | |
139.91 | |
139.91 | |
BF3 45.5% |
Safety and Handling
GHS H Statement
H301-H330-H372-H314-H318-H227
Toxic if swallowed.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P270-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H332-H372
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 207-325-9
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only