Fisherbrand™ Borosilicate Cylinders with Pouring Lip
Heavy-wall construction resists breakage
 View more versions of this product
Catalog No. 08-530L
Explore available promotions
Description
Description
Heavy-wall construction resists breakage
  • Ungraduated
  • Fused round base
  • For use with hydrometers
Specifications
Specifications
20.29 oz.
Round
375 mm
2 in.
600 mL
Round
Ungraduated Cylinder
14.17 in.
51 mm
Borosilicate Glass
Videos
SDS
Documents
Documents
Accessories