Learn More
Eisco™ Borosilicate Salt Bridge
The Borosilicate Salt Bridge is used to connect the oxidation and reduction half-cells of a galvanic cell. It allows ions to flow from one solution to the other without mixing and allows the solutions to remain electrically neutral.
Supplier: Eisco™ CH0744
Avertissement:
This item is intended for educational purposes only.
Description
- Comprises an inverted U-tube with its ends closed by sintered discs.
- For use in experiments involving the measurement of cell EMF under conditions of forward, zero and reverse current flow.
- This apparatus provides a more effective method of making reversible cells then the wet filter paper or agar gel methods.
Spécifications
| U-Shaped Tube | |
| 3.9 in. | |
| 3.1 in. |
| Blood | |
| 100 mm | |
| 80 mm |