missing translation for 'onlineSavingsMsg'
Learn More

Bortezomib, 98%

Bortezomib (CAS # 179324-69-7) is an inhibitor of protein degradation pathways. | CAS: 179324-69-7 | C19H25BN4O4 | 384.243 g/mol

$473.10 - $826.16

Chemical Identifiers

CAS 179324-69-7
Molecular Formula C19H25BN4O4
Molecular Weight (g/mol) 384.243
MDL Number MFCD09056737
InChI Key GXJABQQUPOEUTA-RDJZCZTQSA-N
Synonym bortezomib, velcade, bortezomib ps-341, dpba, pyz-phe-boroleu, unii-69g8bd63pp, velcade tn, bortezomib velcade, 1r-3-methyl-1-2s-1-oxo-3-phenyl-2-pyrazinylcarbonyl amino propyl amino butyl boronic acid, 1r-3-methyl-1-2s-3-phenyl-2-pyrazine-2-carbonylamino propanoyl amino butyl boronic acid
PubChem CID 387447
ChEBI CHEBI:52717
IUPAC Name [(1R)-3-methyl-1-[[(2S)-3-phenyl-2-(pyrazine-2-carbonylamino)propanoyl]amino]butyl]boronic acid
SMILES B(C(CC(C)C)NC(=O)C(CC1=CC=CC=C1)NC(=O)C2=NC=CN=C2)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60378MA
View Documents
Thermo Scientific Chemicals
J60378MA
10 mg
Each for $473.10
Only null left
 
AAJ60378MB
View Documents
Thermo Scientific Chemicals
J60378MB
25 mg
Each for $826.16
Only null left
 
Description

Description

A potent, highly selective and reversible inhibitor of the 20S proteasome, Bortezomib is used as an antineoplastic agent, which controls the growth of cancer cells. It is also useful in the treatment of multiple myeloma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Bortezomib is an antineoplastic compound derived from boronic acid
• Bortezomib reversibly inhibits the 26S proteostome that maintains cell cycle pathways related to growth and division
• Bortezomib is an inhibitor of NF-κB, a factor that is constituitively active in some cancers

Application

• Bortezomib is suitable for use in the research of multiple myeloma and lymphomas, and for use in the research of protein degradation pathways
• Bortezomib is intended for in vitro and biochemical applications

Specifications

Chemical Identifiers

179324-69-7
384.243
GXJABQQUPOEUTA-RDJZCZTQSA-N
387447
[(1R)-3-methyl-1-[[(2S)-3-phenyl-2-(pyrazine-2-carbonylamino)propanoyl]amino]butyl]boronic acid
C19H25BN4O4
MFCD09056737
bortezomib, velcade, bortezomib ps-341, dpba, pyz-phe-boroleu, unii-69g8bd63pp, velcade tn, bortezomib velcade, 1r-3-methyl-1-2s-1-oxo-3-phenyl-2-pyrazinylcarbonyl amino propyl amino butyl boronic acid, 1r-3-methyl-1-2s-3-phenyl-2-pyrazine-2-carbonylamino propanoyl amino butyl boronic acid
CHEBI:52717
B(C(CC(C)C)NC(=O)C(CC1=CC=CC=C1)NC(=O)C2=NC=CN=C2)(O)O

Specifications

179324-69-7
Brown to White
MFCD09056737
UN3249
bortezomib, velcade, bortezomib ps-341, dpba, pyz-phe-boroleu, unii-69g8bd63pp, velcade tn, bortezomib velcade, 1r-3-methyl-1-2s-1-oxo-3-phenyl-2-pyrazinylcarbonyl amino propyl amino butyl boronic acid, 1r-3-methyl-1-2s-3-phenyl-2-pyrazine-2-carbonylamino propanoyl amino butyl boronic acid
GXJABQQUPOEUTA-RDJZCZTQSA-N
[(1R)-3-methyl-1-[[(2S)-3-phenyl-2-(pyrazine-2-carbonylamino)propanoyl]amino]butyl]boronic acid
387447
384.24
Powder
139°C to 143°C
C19H25BN4O4
10 mg
14,1351
Soluble in chloroform,dimethyl sulfoxide,ethanol and methanol.
B(C(CC(C)C)NC(=O)C(CC1=CC=CC=C1)NC(=O)C2=NC=CN=C2)(O)O
384.243
CHEBI:52717
99%
Bortezomib
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H372
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P314-P330-P361-P363-P501c

H301+H311-H330-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MEDICINE, SOLID, TOXIC, N.O.S.

RTECSNumber : ED7771666

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only