BrandTech™ Bottle adapters for BRAND Bottletop Dispensers and Bottletop Burettes
Adapters for common bottle threads and standard taper joints
$36.41 - $213.53
Catalog Number | Mfr. No. | Description | Material | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Material | Price | Quantity | ||||
13-880-715
|
BrandTech™
704375 |
External Thread: 33mm; Bottle Thread: 24 mm | PTFE, ETFE |
Each for $101.06
|
|
||||
13-880-716
|
BrandTech™
704378 |
External Thread: 33 mm; Bottle Thread/Ground Joint: 28mm/S28mm | PTFE, ETFE |
Each for $112.18
|
|
||||
13-880-717
|
BrandTech™
704388 |
External Thread: 33 mm; Bottle Thread: 38 mm | PTFE, ETFE |
Each for $117.56
|
|
||||
13-880-718
|
BrandTech™
704390 |
External Thread: 33 mm; Bottle Thread: 40 mm | PTFE, ETFE |
Each for $129.65
|
|
||||
13-880-719
|
BrandTech™
704391 |
External Thread: 45 mm; Ground Joint: S40 mm | PTFE, ETFE |
Each for $213.53
|
|
||||
13-880-720
|
BrandTech™
704395 |
External Thread: 33 mm; Bottle Thread: 45 mm | PTFE, ETFE |
Each for $129.00
|
|
||||
13-880-721
|
BrandTech™
704398 |
External Thread: 45 mm; Bottle Thread: 33 mm | PTFE, ETFE |
Each for $126.50
|
|
||||
13-880-745
|
BrandTech™
704325 |
External Thread: 33 mm; Bottle Thread: 24mm | Polypropylene |
Each for $36.41
|
|
||||
13-880-746
|
BrandTech™
704340 |
External Thread: 33 mm; Bottle Thread: 40 mm | Polypropylene |
Each for $36.41
|
|
||||
13-880-747
|
BrandTech™
704345 |
External Thread: 33mm; Bottle Thread: 45 mm | Polypropylene |
Each for $36.71
|
|
||||
13-880-748
|
BrandTech™
704343 |
External Thread: 45mm ; Ground Joint: S40 mm | Polypropylene |
Each for $36.41
|
|
||||
13-880-749
|
BrandTech™
704419 |
External Thread: 33 mm; Ground Joint: STJ 19/26 | Polypropylene |
Each for $122.32
|
|
||||
13-880-750
|
BrandTech™
704424 |
External Thread: 33 mm; Ground Joint: STJ 24/29 | Polypropylene |
Each for $121.74
|
|
||||
13-880-751
|
BrandTech™
704429 |
External Thread: 33 mm; Ground Joint: STJ 29/32 | Polypropylene |
Each for $143.21
|
|
||||
Description
- Economical polypropylene adapters work for most applications
- ETFE/PTFE adapters for aggressive acids and solvents
- Leading number refers to male thread of adapter
- Second number refers to bottle neck the adapter fits
- 28mm, 24mm and 22mm bottle threads require stacking adapter with 45/33mm adapter.