George Taylor Brass & Bronze Works Bottle Stopper Remover
Supplier: George Taylor Brass & Bronze Works TBB69210
Description
Remove “frozen” ground-glass stoppers from bottles. Stopper remover rests on bottle mouth. Inner jaws grasp stopper firmly. Stopper is extracted by carefully turning knob. Apparatus is adjustable to fit various bottle sizes.
Specifications
Bottle Stopper Remover | |
Removes frozen ground glass stoppers from bottles |