missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Bottle Top Dispenser with Re-Circulation Valve
Provides a contemporary design coupled with a recirculation valve. Bottle Top Dispenser with Re-Circulation Valve has enhanced chemical compatibility and smooth piston movement.
Description
- Re-circulation Valve helps user in quick purging without wasting any reagent
- Specially designed unique spring-less valve provides greater chemical compatibility, no chemical deposition and minimizing the maintainence
- Presence of a unique spring-less valve, PTFE piston and ETP O-ring provides greater chemical compatibility, ensures a wiper like smooth and effortless piston movement
- Specially designed 180° rotation volume setting knob provides quick and effortless volume adjustment
- Adjustable Delivery Nozzle provides flexibility to the users in demanding laboratory conditions
- Bottle Neck Adapters of different sizes (28, 30, 32, 38, 40 and 45mm) ensures compatibility of dispenser with a majority of reagent bottles
- Each dispenser is accompanied with a tool, which helps in easy and quick re-calibration
- Contemporary and well designed contours, Silky surface finish
- Individually calibrated as per ISO 8655-2
Specifications
Bottletop Dispenser | |
CE Certificate, ISO 13485-2003, ISO 9001-2008, Compliance as per ISO 8655, Calibration lab certification by NABL as per ISO 17025 | |
2.2 g/cm3 | |
Polypropylene - Outer Body (10% Glass Filled) | |
Bottle Top Dispenser is used in laboratories to dispense reagent from the bottle in a preset volume accurately and safely into a receiving vessel. | |
300mbar to 600mbar | |
1 year free replacement warranty against any manufacturing defects. |
Autoclavable | |
White and Fisher Blue | |
Shape: Cylindrical | |
0.5 to 5 mL | |
Calibration tool, Adapters set (5 nos.), Instruction Manual, Inlet tube, Calibration report | |
500mm2 |