missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bouin's Fixative Solution, Picric Acid-Formalin-Acetic Acid Mixture, Ricca Chemical
$186.18 - $467.47
Specifications
Chemical Name or Material | Bouin's Fixative Solution |
---|---|
CAS Min % | 68.0,24.33,4.76,0.87 |
CAS Max % | 71.0,25.33,4.96,0.91 |
CAS | 50-00-0,64-19-7,88-89-1 |
Packaging | Natural Poly Bottle |
Specifications
Bouin's Fixative Solution | |
71.0,25.33,4.96,0.91 | |
Natural Poly Bottle |
68.0,24.33,4.76,0.87 | |
50-00-0,64-19-7,88-89-1 |