missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Br-Mmc (=4-Bromomethyl-7-methoxycoumarin) 98.0+%, TCI America™
Supplier: TCI America A55515G
Specifications
Br-Mmc (=4-Bromomethyl-7-methoxycoumarin) [for HPLC Labeling] | |
215°C | |
C11H9BrO3 | |
5 g | |
4-bromomethyl-7-methoxycoumarin, 4-bromomethyl-7-methoxy-2h-chromen-2-one, br-mmc, ccris 7996, 4-bromomethyl-7-methoxychromen-2-one, 2h-1-benzopyran-2-one, 4-bromomethyl-7-methoxy, 4-bromomethyl-7-methoxy-2-oxo-2h-benzopyran, acmc-1agnd, 4-brommethyl-7-methoxy-2h-chromen-2-on, 4-bromomethyl-7-methoxy coumarin | |
COC1=CC=C2C(CBr)=CC(=O)OC2=C1 | |
269.09 | |
269.09 | |
Crystalline Powder |
35231-44-8 | |
White-Yellow | |
MFCD00006869 | |
3261 | |
CTENSLORRMFPDH-UHFFFAOYSA-N | |
4-(bromomethyl)-7-methoxy-2H-chromen-2-one | |
121894 | |
≥98.0% (T) |
Chemical Identifiers
35231-44-8 | |
269.09 | |
CTENSLORRMFPDH-UHFFFAOYSA-N | |
121894 | |
COC1=CC=C2C(CBr)=CC(=O)OC2=C1 |
C11H9BrO3 | |
MFCD00006869 | |
4-bromomethyl-7-methoxycoumarin, 4-bromomethyl-7-methoxy-2h-chromen-2-one, br-mmc, ccris 7996, 4-bromomethyl-7-methoxychromen-2-one, 2h-1-benzopyran-2-one, 4-bromomethyl-7-methoxy, 4-bromomethyl-7-methoxy-2-oxo-2h-benzopyran, acmc-1agnd, 4-brommethyl-7-methoxy-2h-chromen-2-on, 4-bromomethyl-7-methoxy coumarin | |
4-(bromomethyl)-7-methoxy-2H-chromen-2-one |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator