Learn More
Thermo Scientific Chemicals Bradford Dye Reagent, Ready-to-Use soln.
Suitable for protein concentration determination (100 to 1500μg)
$194.18 - $319.52
Specifications
| Chemical Name or Material | Bradford Dye Reagent |
|---|---|
| Color | Blue |
| Name Note | Ready-to-Use soln. |
| UN Number | UN1805 |
| Solubility Information | Miscible with water. |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ61522AP
|
Thermo Scientific Chemicals
J61522AP |
500 mL |
Each for $194.18
|
|
|||||
|
AAJ61522K2
|
Thermo Scientific Chemicals
J61522K2 |
1 L |
Each for $319.52
|
|
|||||
Description
The Bradford Reagent is suitable for protein concentration determination (100-1500μg).Bradford dye reagent is used in the determination of protein content in cell fractions and assessing the protein concentrations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Bradford Dye Reagent | |
| Ready-to-Use soln. | |
| Miscible with water. |
| Blue | |
| UN1805 | |
| Liquid |
RUO – Research Use Only