Brady™ 55 Gallon Drum Spill Kit with wheels - Chemical
Absorbs up to 38 gal. per kit
Supplier:  Brady™ SKH55W
Catalog No. 19-140-868
Explore available promotions
Includes: 15 x 19 in. pads (60); 3 x 12 in. SOCs(4); 17 x 19 in. pillows (8); One pair nitrile gloves; Diaposal bags (5); Goggles, Emergency Response Handbook.
Description
Description
  • Can be easily opened and closed for fast response to medium sized spill of hazardous materials
  • Dia. x H: 61 x 94 cm (24 x 37 in.)
  • Compliance
    UN specifications
    Specifications
    Specifications
    55 gal.
    38 gal.
    Polyethylene
    55 gal. wheeled drum container, (4) 3 in. x 12 ft. socks, 5 disposal bags, (60) 15 x 19 in. chemical pads, (8) 18 x 18 in. pillows, 1 pair of goggles, handbook, 1 pair of Nitrile gloves
    58.42 cm
    58.42 cm
    124.58 kg
    208 L
    144 L
    Chemical Sorbent
    23 in.
    23 in.
    56.5 lb.