Brady™ Allwik™ 6.5 gal. Spill Bucket, Universal
Accessible spill kit in a convenient reusable bucket
Supplier:  Brady™ SKABKT
Catalog No. 17-111-216
Explore available promotions
Includes: (10) - 15 x 19 in. Pads, (3) - 3 in. x 4 ft. SOCs, (1) - Pair Nitrile Gloves, (2) - Disposal Bags, Instruction Sheet
Description
Description
  • For a small spill response of an oil based material
  • Absorbency: 9 gal/bale
Specifications
Specifications
9 gal./Bale
6.5 gal. Spill Bucket, Universal
8.5 lb.
Polyethylene
(10) 15 x 19 in. pads, (3) 3 in. x 4 ft. SOCs, 1 pair Nitrile gloves, 2 disposal bags, instruction sheet
18.74 kg
Product Suggestions
Product Suggestions