Brady™ Ball Valve Lockout Devices
Fit all major brand ball valves
Supplier:  Brady™ 65666
 View more versions of this product
Catalog No. 17-987-275D
Explore available promotions
Description
Description
  • Lock ball valves to help prevent accidental start-ups
  • Patented, easy-to-use one-piece design
Compliance
Assist in compliance with OSHA's lockout standard.
Specifications
Specifications
Ball Valve Lockout
Nylon
0.8 cm
Valve Size: 0.25 to 1 in. (0.6 to 2.5cm)
Red
OSHA
To help prevent accidental start-ups