Brady™ BBP™ Heavy-Duty Print Ribbon
BBP™ Heavy-Duty B30 Series Drop & Click print ribbons enable 5-second ribbon supply changes. No calibrating, no threading, no waste, no hassles and the ink always faces the right direction.
Supplier:  Brady™ B30R10000GN
 View more versions of this product
Catalog No. 19-211-105
Explore available promotions
Description
Description
  • One-color ribbon for printing a single color onto a label.
  • ''Drop & Click'' print ribbons for quick and easy supply changes. R6000 Series Halogen-Free Resin Formulation offers excellent solvent, scratch and heat resistance.
  • R4300 Series Ribbons offers strong environmental resistance against smearing and chemicals.
  • R4400 Series Resin Formulation offers excellent resistance to solvents, scratches and high heat when printed on the appropriate label material.
  • R10000 series ribbons are based on a resin formulation that offers a high performance image with strong solvent and scratch resistance.
  • Ribbons are tough, smear-proof that produce long-lasting printed text.
  • Print can withstand years in industrial areas and outdoor conditions.
  • No calibrating, no threading, no waste, no hassles.
Specifications
Specifications
Printer Ribbon
BBP30 Label Printer, BBP31 Sign and Label Printer, BBP33 Label Printer, BBP35 Multi-Color Sign and Label Printer, BBP37 Color and Cut Sign and Label Printer
Thermal Transfer
4.3 in.
110 mm
Green Heavy-Duty Print Ribbon for BBP3x Printers
0.5 lb.
For solvent and scratch resistance
B30-R10000-GN
Green
Resin
200 ft.
60.96 m
Non-sterile
Ribbon
0.22 kg
1 Ea.
Product Suggestions
Product Suggestions