Brady™ BBP™ NOTICE Pre-Printed Pre-Cut Blank Sign Headers
BBP™ NOTICE Pre-Printed Pre-Cut Blank Sign Headers are made of durable, low-shrink vinyl with ultra-aggressive adhesive that is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
Supplier:  Brady™ B30241595ANSINO
 View more versions of this product
Catalog No. 19-211-127
Explore available promotions
Description
Description
  • Pre-printed headers result in multicolor header signs using a single, black print pass
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces—including outdoors
  • Conforms and adheres to "difficult" surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
  • Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
Specifications
Specifications
B30-241-595-ANSINO
Gloss
Vinyl
Permanent Acrylic
Pipemarkers, arrow tape, warehouse marking, warning panels, safety signs, arc flash and visual workplace labels
75 mm
-40°F
3 in.
-40°C to 82°C
English
Notice
Blue on White
ASME ANSI Z535, OSHA 1910.145
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
300/Pk.
Pre-Printed Pre-Cut Blank Sign headers for BBP3x Printers
2.25 in.
180°F
0.004 in.
76.2 mm
-40°F to 180 °F
5.7 H x 7.6 cm W
Indoor/Outdoor Tape