missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Blank Hazardous Communication Panels
Blank panels to create customized signs
$48.81 - $267.18
Specifications
Displayed Text | NFPA Diamond |
---|---|
Product Type | Hazardous communication label |
Catalog Number | Mfr. No. | Width (English) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Width (English) | Price | Quantity | |||||
19016238
|
Brady™
58501 |
5 in. |
Pack of 10 Each for $73.47
|
|
|||||
19016239
|
Brady™
BRD58502 |
7.5 in. |
Pack of 10 Each for $126.06
|
|
|||||
19016240
|
Brady™
BRD58503 |
10 in. |
Pack of 10 Each for $267.18
|
|
|||||
19-016237
|
Brady™
BRD58500 |
2.5 in. |
Pack of 10 Each for $48.81
|
|
|||||
19-016313
|
Brady™
58656 |
11 in. |
Pack for $68.00
|
|
|||||
Description
Make signs to identify hazardous chemical containers and storage areas.
- Place directly on storage tanks, processing tanks and vessels, feed hoppers, pipelines, and other potentially hazardous equipment
- Upper three corners of signal rate health, fire and reactivity on a scale of 0 to 4; lower corner provides specific hazard information
- Available in vinyl with permanent pressure-sensitive adhesive or in rigid fiberglass with predrillled holes for mounting to walls
Compliance
Signals meet NFPA Code 704M and Federal Standard 313.
Specifications
NFPA Diamond | |
Hazardous communication label |