Brady™ Caution and Warning Tapes
Various tape designs in choice of two materials
Supplier:  Brady™ 76312
 View more versions of this product
Catalog No. 19-017131
Explore available promotions
Description
Description
  • Ideal for temporary use outdoors or permanent indoor warning
  • Sold as roll
B-957 Reflective Vinyl with Adhesive
  • Thickness: 1.65 to 1.91mm (0.065 to 0.075 in.)
  • Service temp.: -40° to 70°C (-40° to 160°F)
B-950 Encapsulated Vinyl with Adhesive
  • Thickness: 0.15mm (0.006 in.)
  • Service temp.: -40° to 80°C (-40° to 176°F)
Compliance
B-950 warning stripe and check tape meets OSHA specifications.
Specifications
Specifications
Black on Yellow