Brady™ Danger Hazardous Area Signs
Danger: Hazardous Area signs (B-302) are high-performance, pressure-sensitive safety signs constructed of surface-printed polyester with a polyester overlaminate.
Supplier:  Brady™ 85493
 View more versions of this product
Catalog No. 19-823-253
Explore available promotions
Description
Description
  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
  • Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)
Specifications
Specifications
Globally Harmonized System of Classification and Labeling of Chemicals (GHS);NFPA 704;OSHA 1910.1200
Polyester
Permanent Acrylic
7 in.
-40°F
10 in.
-40°C to 110°C
7 x 10 in.
Chemical and Hazardous Materials Sign
Industrial
1 Ea.
Identification of fire alarms, extinguishers and emergency exits
230°F
0.006 to 0.01 in.
25.4 cm
-40°F to 230°F
DANGER Hazardous Area
Red on White