missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Danger Hazardous Area Signs
Danger: Hazardous Area signs (B-302) are high-performance, pressure-sensitive safety signs constructed of surface-printed polyester with a polyester overlaminate.
Supplier: Brady™ 85497
Description
- Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
- Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
- Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
- Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)
Specifications
ANSI Z535, OSHA 1910.145 | |
Polyester | |
Permanent Acrylic | |
Identification of fire alarms, extinguishers and emergency exits | |
230°F | |
0.006 to 0.01 in. | |
25.4 cm | |
-40°F to 230°F | |
DANGER Hazardous Materials | |
Red on White |
Industrial | |
1 Ea. | |
Surface Mount | |
7 in. | |
-40°F | |
10 in. | |
-40°C to 110°C | |
7 x 10 in. | |
Chemical and Hazardous Materials Sign |