Brady™ Fire Fire Extinguisher (V-Style) Signs
Fire Fire Extinguisher (V-Style) Signs are high visibility signs from Brady™ that feature bright colors, graphics and pictograms to effectively communicate a safety message or identify the location of a given item.
Supplier:  Brady™ V1FE15A
 View more versions of this product
Catalog No. 19-807-350
Explore available promotions
Description
Description
  • V-style 45° high visibility signs project from the wall to be easily viewed from all angles
  • The legend is repeated on both outward facing sides of a high visibility V-style sign
  • Used to clearly mark fire safety equipment for quick and easy access in an emergency situation
  • Constructed of rigid acrylic material or polyethylene
Specifications
Specifications
Acrylic
Surface Mount
4 in.
4 x 8 x 6 in.
Standard V Sign
1 Ea.
6 in.
203.2 mm
Fire Extinguisher
White on Red