Learn More
Brady™ Floor Safety Signs
Durable, highly visible Floor Safety Signs are designed to warn of hazardous areas, putting valuable safety information at the point of need.
Supplier:  Brady™ 49067
 View more versions of this product
Catalog No. 19-054-840
Only null left
Explore available promotions
Description
Description
  • Floor safety signs improve facility safety by alerting employees to hazards and/or important safety information in areas with foot traffic
  • B-819 Anti-Skid Vinyl features a velvet embossed rigid overlaminate
  • Excellent chemical, oil and water resistance
  • Minimum application temperature of 0°F (-18°C)
Specifications
Specifications
ASTM 4.2.1
1 Ea.
17 in.
Designed to warn of hazardous areas, putting valuable safety information at the point of need
-40°F
-40°C to 70°C
Emergency Exit
Red on White
Vinyl
Permanent Acrylic
Surface Mount
158°F
0.01 in.
-40°F to 158°F
Floor Safety Sign
Product Suggestions
Product Suggestions