missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Floor Safety Signs
Durable, highly visible Floor Safety Signs are designed to warn of hazardous areas, putting valuable safety information at the point of need.
Supplier: Brady™ 92404
Description
- Floor safety signs improve facility safety by alerting employees to hazards and/or important safety information in areas with foot traffic
- B-819 Anti-Skid Vinyl features a velvet embossed rigid overlaminate
- Excellent chemical, oil and water resistance
- Minimum application temperature of 0°F (-18°C)
Specifications
| Abrasion Resistance, Method 5306 of U.S. Federal Test Method Std. No. 191A | |
| 1 Ea. | |
| 17 in. | |
| 158°F | |
| 0.01 in. | |
| -40°F to 158°F | |
| Floor Safety Sign |
| Vinyl | |
| Permanent Acrylic | |
| Designed to warn of hazardous areas, putting valuable safety information at the point of need | |
| -40°F | |
| -40°C to 70°C | |
| Watch Out For Forklift Trucks | |
| Black on Yellow |