Brady™ Floor Safety Signs
Durable, highly visible Floor Safety Signs are designed to warn of hazardous areas, putting valuable safety information at the point of need.
Supplier:  Brady™ 92405
 View more versions of this product
Catalog No. 19-054-889
Explore available promotions
Description
Description
  • Floor safety signs improve facility safety by alerting employees to hazards and/or important safety information in areas with foot traffic
  • B-819 Anti-Skid Vinyl features a velvet embossed rigid overlaminate
  • Excellent chemical, oil and water resistance
  • Minimum application temperature of 0°F (-18°C)
Specifications
Specifications
Abrasion Resistance, Method 5306 of U.S. Federal Test Method Std. No. 191A
1 Ea.
17 in.
Designed to warn of hazardous areas, putting valuable safety information at the point of need
-40°F
-40°C to 70°C
Ear Protection Area
Black on Yellow
Vinyl
Permanent Acrylic
Floor Mount
158°F
0.01 in.
-40°F to 158°F
Floor Safety Sign