missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Globally Harmonized System (GHS) Explosive Picto Labels
For explosive, self-reactive, organic peroxides
$10.42 - $13.66
Specifications
| Color | Black, Red, White |
|---|---|
| Product Type | Pictogram Label |
| For Use With (Application) | Explosives, Self-reactives, Organic Peroxides |
| Catalog Number | Mfr. No. | Quantity | Material | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Material | Price | Quantity | ||||
|
19-140-512
|
Brady™
118815 |
39/Pk. | Vinyl |
Pack of 39 Each for $13.66
|
|
||||
|
19-140-521
|
Brady™
118869 |
39/Pk. | Coated Paper |
Pack of 39 Each for $10.42
|
|
||||
|
19-140-530
|
Brady™
118816 |
16/Pk. | Vinyl |
Pack of 16 Each for $13.23
|
|
||||
|
19-140-539
|
Brady™
118870 |
16/Pk. | Coated Paper |
Pack of 16 Each for $10.42
|
|
||||
|
19-140-548
|
Brady™
118817 |
4/Pk. | Vinyl |
Pack of 4 Each for $13.23
|
|
||||
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards.
- Legend: Explosive
- Made of Vinyl or Coated paper
- Available in three sizes
Specifications
| Black, Red, White | |
| Pictogram Label | |
| Explosives, Self-reactives, Organic Peroxides |