Brady™ GlobalMark™ Label Printer: Grommets
GlobalMark™ Label Printer: Grommets are used to reinforce a round, pre-punched hole in a safety tag printed on a Brady™ Thermal Transfer Printer.
Supplier: Brady™ BRD20596
Description
- Grommets extend the life of data tags by preventing tearing and abrasion
- Black plastic grommets snap in easily to the pre-punched holes in thermal transfer printable tags from Brady™
- Allows tags to be customized or serially printed in a continuous manner on a thermal transfer printer before the grommet is applied
- Package includes grommets only (printer and tags sold separately)
Specifications
Grommets | |
Non-sterile | |
Industrial label maker products | |
100/Pk. |