Brady™ Key Retaining Steel Padlocks
Key-Retaining Padlocks prevent the key from being removed until the padlock is fully closed. Available in laminated steel with 2 in. clearance hardened steel shackles to withstand severe physical abuse.
Supplier:  Brady™ 118979
 View more versions of this product
Catalog No. 19-140-383
Explore available promotions
Description
Description
  • Reinforced laminated steel withstands sever physical abuse while hardened steel shackle resists tampering
  • Body plates individually coated for superior rust protection
  • 5-pin cylinder offers more unique key cuts for better tamper resistance
  • Options: keyed differently or keyed alike
Specifications
Specifications
FDA, RoHs, ANSI Z244, OSHA 29 CFR 1910.147
2 in.
5 cm
White
Keyed Alike
5-pin Cylinder
2 in. Shackle Steel Lock
Key-retaining Padlock
1.2 in.
3.33 cm
1.6 in.
1 in.
0.25 in.
0.6 cm
2.54 cm
3 cm
Steel
For Safety Lockout Purposes
Outdoor; Industrial Manufacturing; Harsh Environments