Brady™ NFPA Placard Systems
Customize placards on the spot with NFPA squares blue, white, red and yellow
Supplier:  Brady™ 50053
 View more versions of this product
Catalog No. 19-807-075
Explore available promotions
Description
Description
  • Available in fiberglass or vinyl
  • Includes 10 blank placards and 10 characters each of numbers 0-4, symbols for Use No Water, Radiation, OXY, ACID, COR, and ALK
Specifications
Specifications
10 Blank placard and 10 character each of number 0 to 4, symbol (For Use No Water, Radiation, Oxidizer, ACID, Corrosive and Alkali)
1 Ea.
180°F
0.004 in.
-40°F to 180°F
NFPA Diamond Placard
Yellow on White
Vinyl
Pipemarkers, arrow tape, number and letter, vehicle placard, pedestal marker, warning panel and safety sign
-40°F
-40°C to 82°C
25.4cm (10 in.) square
NFPA Placard Kit
Product Suggestions
Product Suggestions