Brady™ ToughStripe™ Floor Marking Corner Marks
Durable tape perfect for marking skid and pallet storage areas, as well as other designated areas
Supplier:  Brady™ 104430
 View more versions of this product
Catalog No. 19150760
Explore available promotions
Description
Description
Durability
  • Withstands forklift traffic without tearing or lifting
  • Low-profile design minimizes tears and scratches from skids, pallet jacks, etc.
  • Made with rugged polyester and aggressive adhesive
Painless Removal
  • Does not chip and flake when being removed - comes off in one piece
  • Clean removal without any unsightly adhesive residue or damage to the floor
High Visibility
  • High-gloss surface shines like new paint
  • Low profile minimizes debris buildup along the edges that occurs with thicker (extruded) floor marks
  • Surface resists marks and smudges and cleans up like new
  • Available in a variety of high-impact colors
Specifications
Specifications
TS-50.80-514-COR-RD-20
ToughStripe Diecut Shapes (L's, T's, +'s, footprints and more)
Red
Polyester
115 Gardner Units
L-shaped
-18°C to +54°C
0.201 mm
12.7 cm
50.80 mm
ANSI A1264.2 2006, OSHA 1910.22
Rubber, Permanent
Tape
For floor corner mark, Used to mark aisles, passageways, storage locations and point out safety equipment and exits
20/Pk.
0°F to 130°F
0.008 in.
5 in.
2 in.