missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ Bradycone™ Warning System - 35 in H

Use our Bradycone™ Warning System for any of your hazardous warning needs

$142.96 - $142.96

Specifications

Color Yellow
Height (English) 35 in.
Packaging Quantity 1 Ea.
Width (Metric) 323.85 mm
View More Specs

Products 4
Catalog Number Mfr. No. Displayed Text Price Quantity  
Catalog Number Mfr. No. Displayed Text Price Quantity  
19-104-399
View Documents
Brady™
103680
CAUTION/PRECAUCION WET FLOOR/PISO MOJADO
Each for $142.96
 
19-104-400
View Documents
Brady™
103681
Each for $142.96
 
19-104-401
View Documents
Brady™
103682
Each for $142.96
 
19-104-402
View Documents
Brady™
103683
Each for $142.96
 
Description

Description

  • Cones are four-sided, so your message can't be missed
Specifications

Specifications

Yellow
35 in.
1 Ea.
323.85 mm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'