missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BrandTech™ BRAND™ Four-Clear Sided Disposable Cuvettes for Fluorescence
German manufactured premium cuvettes
Supplier: BrandTech™ BRAND™ 759125
Description
- BRAND premium quality four sided clear disposable cuvettes for best results in fluorescence applications.
- Choose polystyrene for general purpose requirements in the visible range (340-800nm).
- Choose proprietary UV-plastic cuvettes with virtually no autoflourescence for best signal to noise ratio for assays in the 230-800nm range.
- 10mm lightpath to fit most fluorimeters.
Specifications
Fluorimetry Cuvette | |
Single-use | |
UV-transparent Plastic | |
100 | |
230 to 800 nm | |
flourometers | |
Fluorimetry |
BRAND four sided clear cuvettes for fluorimetry. Proprietary UV-plastic. Package of 100 | |
2.5 to 4.5 mL | |
12.5mm | |
10 mm | |
45 mm | |
12.5 mm |