BrandTech™ BRAND™ BLAUBRAND™ Glass Volumetric Flasks, Class A, USP, Certified
Indispensable for preparing dilutions and standard solutions
Supplier:  BrandTech™ BRAND™ 36977
 View more versions of this product
Catalog No. 13-690-130
Explore available promotions
Description
Description
  • Includes with glass stopper with dripping tip
  • Contrast blue enamel print on transparent flasks ensures optimum combination of resistance and readability
  • Available in all ISO sizes from 5 to 2000mL
  • Available in transparent and amber colors
  • Made of low expansion coefficient, chemically resistant borosilicate 3.3 glass
  • Supplied with batch number and an accompanying USP batch certificate in original packaging
Specifications
Specifications
USP
25 mL
A
Transparent
Glass
NS 10/19