BrandTech™ BRAND™ Plastic Spectrophotometry Cuvettes
BRAND™ Plastic Spectrophotometry Cuvettes with mold-cavity matched for the lowest variability in extinction coefficient.
Supplier:  BrandTech™ 759085D
 View more versions of this product
Catalog No. 14-377-016
Explore available promotions
Description
Description
BRAND spectrophotometry cuvettes. Mold-cavity matched for the lowest variability in extinction coefficient.
  • Mold cavity sorted spectrophotometry cuvettes with 2 clear windows
Specifications
Specifications
Non-sterile
Semi-micro
Single-use
PMMA
Life Science Quality
10 mm
45 mm
12.5 mm
10 mm
Spectrophotometer Cuvet
Clear
3 mL
Arrow mark to indicate direction of transmission and reduce variation
500 Pk.
300 nm
Spectrophotometers, Photometers
Spectrophotometry
Rectangle