missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BRAND™ BRANDplates™ pureGrade™ 96-Well Microplates
Constructed of polystyrene with nontreated surface. BRANDplates™ pureGrade™ 96-Well Microplates are noncytotoxic, and free from endotoxins, DNase, DNA, and RNase. Medium binding, nonsterile microplates are suited for homogenous assays, screening, and storage with gray alphanumeric embossing on white and black solid plates for easy well identification.
Description
- Polystyrene surface
- Medium binding, nonsterile
- Standard plate for most applications
- Gray alphanumeric embossing on white and black solid plates for easy well identification
- Strip plates available
- Standard plates available in U-, V-, F-, C-shapes
- Transparent bottoms available in F- shape
- Available in clear, white or black
- Free from endotoxins, DNase, DNA, RNase; noncytotoxic
- Suited for homogenous assays, screening, and storage.
Specifications
Includes Barcode | |
Standard | |
8 x 12 | |
Black | |
350 μL | |
Non-sterile | |
Flat Bottom (F-well) | |
Homogenous assays, screenings and storage | |
Yes |
ANSI, SLAS | |
Without Lid | |
Medium Binding | |
Polystyrene | |
96 | |
Non-treated | |
Yes | |
100/Cs. |