BrandTech™ Low Density Polyethylene Dropping Bottles
VITLAB Dropping bottles are made of low density polyethylene
Supplier:  BrandTech™ 94587
 View more versions of this product
Catalog No. 14-379-055
Explore available promotions
Description
Description
  • Screw caps made of high density polyethylene.
  • Trim top to customize orifice size
  • Push-on cap helps secure contents
Specifications
Specifications
1.6 oz., 50mL
1.6 oz.
GL 18
Screw Cap
50 mL
LDPE
37 mm