BrandTech™ BRAND™ Polypropylene Thread Adapters for Bottle-Top Dispensers
Adapt current model Dispensette™ seripettor™, Titrette™ and QuikSip™ to common bottle sizes.
Supplier:  BrandTech™ BRAND™ 704397
 View more versions of this product
Catalog No. 13-688-321
Explore available promotions
Description
Description
  • Polypropylene Construction
  • For use with Dispensette™ seripettor™ Titrette™ and QuikSip™ Bottle-top dispensers
Specifications
Specifications
Bottle Thread Adapter
Polypropylene
Dispensette™, seripettor™, Titrette™ and QuikSip™
GL 45/38