BrandTech™ Support Jacks
Available in anodized aluminum (blue) or epoxy-powder-coated aluminum (green) decks
Supplier:  BrandTech™ B11015
 View more versions of this product
Catalog No. 03-840-146
Explore available promotions
Description
Description
  • Lift mechanism is made of 304 stainless steel with POM rollers for long service life
Specifications
Specifications
Epoxy-powder Coated Aluminum
2.16 to 4.72 in.
10 cm
10 cm
Support Jack
5.5 to 12 cm
3.93 in.
3.93 in.
22 lb.
Fume hoods and marine and industrial laboratories, POM rollers
Product Suggestions
Product Suggestions