BrandTech™ VACUUBRAND™ Rubber Tubing for Oil-Free Diaphragm Vacuum Pumps
For VACUUBRAND and Diaphragm pumps, and other pumps having a 10mm (3/8 in.) hose barb
Supplier:  BrandTech™ VACUUBRAND™ 20686002M3
Catalog No. 13-878-111
Explore available promotions
Description
Description
  • Inner diameter 10mm
  • Three meter length
  • Thick walls resist collapsing
  • Made of high-quality caoutchouc rubber
Specifications
Specifications
Caoutchouc Rubber
BrandTech pump models: ME1C, MEC, MZ1C, ME 2 and ME 4 NT Pumps
3-meter length