BrandTech™ VACUUBRAND™ VHCpro Suction Handset
Ergonomic light weight suction handset
Supplier: BrandTech™ VACUUBRAND™ 20688061
Description
- Autoclavable at 121°C and 2 barsv
- Ready for use with 2.5 m flow through and flexible silicon tube, adapters for graduated, aspiration and pasteur pipettes, adapter for pipette tips, wall holder for standby-position and clips with adhesive surface to guide the suction tube into the biosafety cabinet neatly
- Permanent suction mode for continuous flow
- Mechanism of the VHCpro handset is not in contact with contaminated fluids and enables reliable, leak free working
Specifications
VHCpro Suction Handset |