Branson Ultrasonics™ 200- and 400-Watt Sonifiers: Horns
Various horns for Branson 200- and 400-Watt Sonifiers
Supplier: Branson Ultrasonics™ 623004020
Description
For use with Branson 200- and 400-Watt Sonifiers
Tapped Horns
- High-gain “Q” horn maintains intensity when processing with large tip diameters
- 1/2 in. horns supplied with one replaceable tip
- Replace only the flat tip to remedy cavitation erosion; use extender with 0.5 in. horns for environmental and other particulate samples
- Transparent polycarbonate water bath with 1/8 NPT flow ports
Specifications
Solid Horn | |
200 to 500 mL | |
Model 500 ultrasonic dismembrator |