Learn More
Branson Ultrasonics™ Bransonic™ General Purpose Cleaner
For ultrasonic cleaners
Supplier: Branson Ultrasonics™ CPN955007
Specifications
| Powder | |
| General Purpose Cleaner: Powder | |
| 2 lb. |
| Used to remove general soils, fingerprints, dust, light oils, greases, Safe on metals, Glass, Plastic | |
| 2 lb. |