Branson Ultrasonics™ DHA-1000 Parts Basket
Basket for DHA1000 to prevent parts from sitting on tank bottom.
Supplier:  Branson Ultrasonics™ CPN916032
Catalog No. 15-335-73
Explore available promotions
Description
Description
  • Basket for DHA1000
  • All 304 stainless steel construction makes baskets compatible with most cleaning processes
  • Type 304 stainless steel is acceptable for use with all alkaline solutions and many acids
  • Basket has Electro polished finish
  • Carryover and dragout of cleaning solution are minimized by open design and construction
Specifications
Specifications
Mesh Basket
Ultrasonic Cleaner, DHA-1000