missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Branson Ultrasonics™ Sonifier™ Continuous Flow Attachment for Models 250, 350, and 450

Processes low-viscosity solutions at rates up to 30lph

Supplier:  Branson Ultrasonics™ 100146171

Catalog No. 22-309792


Explore available promotions

Description

Description

Can be used to emulsify, disperse, and homog-enize by pumping a solution through a zone of intense ultrasonic activity.

  • Degree of processing is controlled by varying the amplitude of the ultra-sonic horn and the flow rate
  • Some solutions may require recirculating until the desired results are obtained
  • Incorporates a cooling jacket through whicha suitable cooling liquid can be circulated to retard heat build-up during extended operation
  • May also be sealed in a closed system to assure sterile conditions and inhibit contamination when working with infectious materials
  • Overall dimension (L x Dia.): 12.7 x 9.65cm (5 x 3.8 in.)

  • Production of vaccines and antigens
  • Emulsification of immiscible liquids (with or w/o surfactants)
  • Removal of cell walls
  • Dispersing metal oxides in solvents

Specifications

Specifications

Continuous Flow Attachment
9.65 cm
Emulsify, disperse and homogenize by pumping a solution through a zone of intense ultrasonic activity
5 in.
Stainless Steel
2.26 kg
3.8 in.
Ultrasonic Cell Disruptor
Cooling jacket
12.7 cm
5 lb.
Videos
SDS
Documents

Documents

Promotions

Promotions