Branson Ultrasonics™ Soundproof Enclosure
For use with Sonifier™ Cell Disruptors
Supplier:  Branson Ultrasonics™ 101063275
Catalog No. 22-309-803
Explore available promotions
Requires : Adapter 15-335-300 (only with 150 watt Sonifier)
Description
Description
For use with Sonifier™ Cell Disruptors
Minimizes mechanical noise produced by ultrasonic processing.
  • Lowers noise levels by 20-25dBA
  • Sturdy cabinet is lined with waterproof, sound-abating material that is impervious to most solutions and reagents and cleans easily
  • See-through door allows process viewing
  • Opens at top and bottom for easy access with a cup horn or standard probes
Order Info
Interior dimensions: 10.5L x12.75W x 20.75 in. H (26.7 x 32.4 x 52.7cm). Overall dimensions: 12L x 14.5W x 23 in. H (30.5 x 36.8 x 58.4cm). Net Weight: 19.5 lb. (8.8kg).
Specifications
Specifications
Soundproof Enclosure
28 in.
12.5 in.
18.5 in.
Ideal for minimizes mechanical noise levels by 20 to 25dBA produced by ultrasonic processing
12.5 x 18.5 x 28 in. (31.8 x 47 x 71.1 cm)
71.1 cm
31.8 cm
47 cm
Sonifier™ cell disruptor