3M™ Breathe Easy™ Complete PAPR Assembly
3M™ Breathe Easy™ Complete PAPR Assembly contains all PAPR parts needed for use in First Responder settings.
Supplier: 3M™ 7100144937
Description
- Complete PAPR Assembly
- Easy to Use
- Works with 3M™ Versaflo™ Standard Battery TR-630 providing longer run time than the 3M™ Breathe Easy™ Turbo Powered Air Purifying Respirator (PAPR) NiMH Battery Pack BP-15
- Comfortable
- 3M Catalog# RBE-10BR
Specifications
5.5 to 13 hours | |
NIOSH Approved | |
Blower motor, butyl rubber hood, breathing tube, decon belt, cartridge, battery adapter, standard battery, easy clean holster, battery charger, cartridge shower cover, and an airflow indicator | |
Back-mounted |
High Capacity Lithium Ion | |
PAPR System with Hood | |
Breathe Easy™ | |
1 Ea. |