missing translation for 'onlineSavingsMsg'
Learn More

Brij™ 30, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 216720025

Catalog No. AC216720025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Brij™ 30
2mg KOH/g max.
2.5kg
145 to 165mg KOH/g
Glass bottle
0.95
C20H42O5
C12H25(OCH2CH2)4OH
01,892
Polyoxyethylene(4)lauryl ether
362.54
9002-92-0
0.9500g/mL
>149°C
Authentic
1.4510 to 1.4530
30 mPa.s (25°C)
1% max.
MFCD00043063
15, 7675
Solubility in water: insoluble. Other solubilities: soluble in lower alcohols, propylene glycol, insoluble in mineral oil, cottonseed oil
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

RTECSNumber : MD0875000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only