missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ BRIJ™35 Detergent, 30% Aqueous Solution, Calbiochem™,
$77.40 - $77.40
Chemical Identifiers
CAS | 9002-92-0 |
---|---|
Molecular Formula | C14H30O2 |
Molecular Weight (g/mol) | 230.39 |
MDL Number | MFCD00043063 |
InChI Key | SFNALCNOMXIBKG-UHFFFAOYSA-N |
Synonym | Polyoxyethyleneglycol Dodecyl Ether |
IUPAC Name | 2-(dodecyloxy)ethan-1-ol |
SMILES | CCCCCCCCCCCCOCCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
203724100ML
|
MilliporeSigma™
203724100ML |
100 mL | Glass Bottle |
Each for $77.40
|
|
||||
Chemical Identifiers
9002-92-0 | |
230.39 | |
SFNALCNOMXIBKG-UHFFFAOYSA-N | |
2-(dodecyloxy)ethan-1-ol |
C14H30O2 | |
MFCD00043063 | |
Polyoxyethyleneglycol Dodecyl Ether | |
CCCCCCCCCCCCOCCO |
Specifications
Undesignated | |
100 mL | |
C14H30O2 | |
Polyoxyethyleneglycol Dodecyl Ether | |
CCCCCCCCCCCCOCCO | |
230.39 |
9002-92-0 | |
Glass Bottle | |
MFCD00043063 | |
SFNALCNOMXIBKG-UHFFFAOYSA-N | |
2-(dodecyloxy)ethan-1-ol |